Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119961 Supplier Invitrogen™ Supplier No. PA5119961
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Raji cell lysate, Jurkat cell lysate, mouse testis tissue lysates, human placenta tissue, human small intestine tissue, human uterus tissue. Subcellular Location: Nucleus.

PHF11 contains two PHD zinc fingers and probably regulates transcription. Distinctive splice variants were expressed in immune tissues and cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF11
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PHF11
Gene Accession No. A6H5X4, Q8BVM9, Q9UIL8
Gene Alias 4933417L10Rik; APY; atopic; BCAP; BRCA1 C-terminus-associated protein; BRCA1-C terminus associated protein; D14Ertd668e; IgE responsiveness; IgE responsiveness (atopic); IGEL; IGER; IGHER; MGC137849 protein; NYREN34; NY-REN-34; NY-REN-34 antigen; PHD finger protein 11; PHD finger protein 11 family member; PHD finger protein 11A; PHD finger protein 11D; PHD finger protein 11-like; Phf11; Phf11-4; Phf11a; Phf11d; Phf11l; Renal carcinoma antigen NY-REN-34; RP11-185C18.3
Gene Symbols PHF11, Phf11a, Phf11d
Host Species Rabbit
Immunogen Recombinant protein within human PHF11 aa 132-331.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219131, 219132, 51131
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.