Learn More
Invitrogen™ PHF11 Polyclonal Antibody

Description
Immunogen sequence: MAQASPPRPE RVLGASSPEA RPAQEALLLP TGVFQVAEKM EKRTCALCPK DVEYNVLYFA QSENIAAHEN CLLYSSGLVE CEDQDPLNPD RSFDVESVKK EIQRGRKLKC KFCHKRGATV GCDLKNCNKN YHFFCAKKDD AVPQSDGVRG IYKLLCQQHA QFPIIAQSAK FSGVKRKRGR KKPLSGNHVQ PPETMKCNTF IRQVKEEHGR HTDATVKVPF LKKCKEAGLL NYLLEEILDK VHSIPEKLMD ETTSESDYEE IGSALFDCRL FEDTFVNFQA AIEKKIHASQ QR; Positive Samples: MCF7, SW480, Raji, Mouse spleen, Mouse thymus, Rat spleen; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | PHF11 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PHF11 |
| Gene Accession No. | A6H5X4, Q5I0E2, Q8BVM9, Q9UIL8 |
| Gene Alias | 4933417L10Rik; APY; atopic; BCAP; BRCA1 C-terminus-associated protein; BRCA1-C terminus associated protein; D14Ertd668e; IgE responsiveness; IgE responsiveness (atopic); IGEL; IGER; IGHER; MGC137849 protein; NYREN34; NY-REN-34; NY-REN-34 antigen; PHD finger protein 11; PHD finger protein 11 family member; PHD finger protein 11A; PHD finger protein 11D; PHD finger protein 11-like; Phf11; Phf11-4; Phf11a; Phf11d; Phf11l; Renal carcinoma antigen NY-REN-34; RP11-185C18.3 |
| Gene Symbols | PHF11, Phf11a, Phf11d |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.