Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 108981AP150UL
Description
PHF11 contains two PHD zinc fingers and probably regulates transcription. Distinctive splice variants were expressed in immune tissues and cells.Specifications
| PHF11 | |
| Polyclonal | |
| Unconjugated | |
| PHF11 | |
| APY, IGEL, IGER, IGHER, NY REN 34, NYREN34, PHD finger protein 11, PHF11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219132, 51131 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A6H5X4, Q9UIL8 | |
| PHF11 | |
| PHF11 Fusion Protein Ag1318 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction