Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHF11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564967
Description
Immunogen sequence: LYSSGLVECED QDPLNPDRSF DVESVKKEIQ RGRKLKCKFC HKRGATVGCD LKNCNKNYHF FCAKKDDAVP QSDGVRGIYK LLCQQHAQFP IIAQSAKFSG VKRKRGRKKP LS Highest antigen sequence identity to the following orthologs - mouse 59%, rat 57%.
PHF11 contains two PHD zinc fingers and probably regulates transcription. Distinctive splice variants were expressed in immune tissues and cells.
Specifications
PHF11 | |
Polyclonal | |
Unconjugated | |
PHF11 | |
4933417L10Rik; APY; atopic; BCAP; BRCA1 C-terminus-associated protein; BRCA1-C terminus associated protein; D14Ertd668e; IgE responsiveness; IgE responsiveness (atopic); IGEL; IGER; IGHER; MGC137849 protein; NYREN34; NY-REN-34; NY-REN-34 antigen; PHD finger protein 11; PHD finger protein 11 family member; PHD finger protein 11A; PHD finger protein 11D; PHD finger protein 11-like; Phf11; Phf11-4; Phf11a; Phf11d; Phf11l; Renal carcinoma antigen NY-REN-34; RP11-185C18.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51131 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIL8 | |
PHF11 | |
Recombinant Human PHF11. Recombinant protein control fragment (Product #RP-105738). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction