Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554400 Supplier Invitrogen™ Supplier No. PA554400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGAVNMCYRT LYIGTGADMD VCLTNYGHCN YVSGKHACIF YDENTKHYEL LNYSEHGTTV DNVLYSCDFS EKTPPTPPSS IVAKVQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Acts as a transcriptional repressor. Involved in recruitment of functional SIN3A complexes to DNA. Represses transcription at least in part through the activity of an associated histone deacetylase (HDAC). May also repress transcription in a SIN3A-independent manner through recruitment of functional TLE5 complexes to DNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF12
Gene Accession No. Q96QT6
Gene Alias 2410142K10Rik; F630045O13; KIAA1523; mKIAA1523; Pf1; PHD factor 1; PHD finger protein 12; PHD zinc finger transcription factor; phf12; PHF12 (HGNC:20816)
Gene Symbols PHF12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF12. Recombinant protein control fragment (Product #RP-93233).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.