Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF13 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297688JF549
Description
PHF13 Polyclonal specifically detects PHF13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PHF13 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC43399, PHD finger protein 13, PHD zinc finger protein PHF5, PHF5, SPOC1, Survival time-associated PHD finger protein in ovarian cancer 1, survival time-associated PHD protein in ovarian cancer | |
Produced in rabbits immunized with E. coli-derived Human PHF13 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
148479 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction