Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555431 Supplier Invitrogen™ Supplier No. PA555431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKRTIVRQGK QVVFRDEDST GNDEDIMVDS DDDSWDLVTC FCMKPFAGRP MIECNECHT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PHF13 contains 1 PHD-type zinc finger. The function of PHF13 is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF13
Gene Accession No. Q86YI8
Gene Alias BC029632; PHD finger protein 13; PHD zinc finger protein PHF5; PHF13; PHF5; SPOC1; Survival time-associated PHD finger protein in ovarian cancer 1; survival time-associated PHD protein in ovarian cancer
Gene Symbols PHF13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF13. Recombinant protein control fragment (Product #RP-90734).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.