Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551406 Supplier Invitrogen™ Supplier No. PA551406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQKEGLLSEA AAEEDIADPF FAYCKQHADR LDRKWKRKNY LALQSYCKMS LQEREKQLSP EAQARINARL QQYRAKAELA RSTRPQAWVP REKLPRPLTS SASAIRKLMR KAELMGISTD IF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

PHF14 gene ontology annotations related to this gene include metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF14
Gene Accession No. O94880
Gene Alias 1110001C23Rik; 4932409F11Rik; 5730446A07Rik; AA623952; AV297001; KIAA0783; mKIAA0783; PHD finger protein 14; Phf14; RGD1563764
Gene Symbols PHF14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF14. Recombinant protein control fragment (Product #RP-93712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.