Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555275 Supplier Invitrogen™ Supplier No. PA555275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASVAADSDVQ VLGPAASPKP LGRLRPPRES KVTRRLPGAR PDAGMGPPSA VAERPKVSLH FDTETDGYFS DGEMSDSDVE AEDGGVQRGP REAGAEEVVR MGVLA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

Scaffold subunit of some HBO1 complexes, which have a histone H4 acetyltransferase activity (PubMed:16387653). Acts as a E3 ubiquitin-protein ligase mediating the ubiquitination and subsequent proteasomal degradation of target protein histone demethylase KDM1A (PubMed:25018020). Also acts as a ubiquitin ligase E3 toward itself. Positive regulator of neurogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF15
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JADE2
Gene Accession No. Q9NQC1
Gene Alias 1200017K05Rik; AI480685; E3 ubiquitin-protein ligase Jade-2; jade family PHD finger 2; jade family PHD finger protein 2; JADE2; JADE-2; Kiaa0239; mKIAA0239; PHD finger protein 15; Phf15; protein Jade-2; protein Jade-2; E3 ubiquitin-protein ligase Jade-2; si:ch73-202g8.3
Gene Symbols JADE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF15. Recombinant protein control fragment (Product #RP-93570).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.