Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF15 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA540976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540976 Supplier Thermo Scientific Supplier No. PA540976

Rabbit Polyclonal Antibody

Peptide sequence: DLAEALVDFI YQYWKLKRKA NANQPLLTPK TDEVDNLAQQ EQDVLYRRLK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

PHF15 Polyclonal antibody specifically detects PHF15 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene JADE2
Gene Accession No. Q9NQC1
Gene Alias 1200017K05Rik; AI480685; E3 ubiquitin-protein ligase Jade-2; jade family PHD finger 2; jade family PHD finger protein 2; JADE2; JADE-2; Kiaa0239; mKIAA0239; PHD finger protein 15; Phf15; protein Jade-2; protein Jade-2; E3 ubiquitin-protein ligase Jade-2; si:ch73-202g8.3
Gene Symbols JADE2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PHF15
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23338
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.