Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF16 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204021H
Description
PHF16 Polyclonal antibody specifically detects PHF16 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PHF16 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
MGC138748, MGC138749, PHD finger protein 16KIAA0215JADE3, protein Jade-3 | |
The immunogen recognized by this antibody maps to a region between residue 773 and 823 of human PHD Finger Protein 16 using the numbering given in entry NP_055550.1 (GeneID 9767). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9767 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction