Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528533 Supplier Invitrogen™ Supplier No. PA528533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene JADE3
Gene Accession No. Q92613
Gene Alias 5730598B06; AI851988; AW544806; jade family PHD finger 3; jade family PHD finger protein 3; JADE3; JADE-3; KIAA0215; mKIAA0215; PHD finger protein 16; PHF16; Protein Jade-3
Gene Symbols JADE3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PHF16. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.