Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145034 Supplier Invitrogen™ Supplier No. PA5145034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGLEKTFAEA RLISAQQKNG VVMPDHGKRR DNRFHCDLIK GDLKDKSFKQ SHKPLRSTDV SQRHLDNTRA ATSPGVGQSA PGTRKEIVPK CNGSLIKVNY NQ Antigen sequence identity: Mouse 89%; Rat 89%.

Scaffold subunit of some HBO1 complexes, which have a histone H4 acetyltransferase activity (PubMed:16387653, PubMed:19187766, PubMed:20129055, PubMed:24065767). Plays a key role in HBO1 complex by directing KAT7/HBO1 specificity towards histone H4 acetylation (H4K5ac, H4K8ac and H4K12ac), regulating DNA replication initiation, regulating DNA replication initiation (PubMed:20129055, PubMed:24065767). May also promote acetylation of nucleosomal histone H4 by KAT5 (PubMed:15502158). Promotes apoptosis (PubMed:16046545). May act as a renal tumor suppressor (PubMed:16046545). Negatively regulates canonical Wnt signaling; at least in part, cooperates with NPHP4 in this function (PubMed:22654112). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF17
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene JADE1
Gene Accession No. Q6IE81
Gene Alias AU041499; D530048A03Rik; gene for apoptosis and differentiation in epithelia; jade family PHD finger 1; jade family PHD finger protein 1; Jade1; Jade1 PHD finger protein; KIAA1807; mKIAA1807; PHD finger protein 17; PHD protein Jade-1; Phf17; Protein Jade-1
Gene Symbols JADE1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 631-732. Recombinant protein control fragment (Product #RP-109729).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.