Learn More
PHF17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA530831
Description
PA5-30831 targets PHF17 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-30831 immunogen is recombit fragment corresponding to a region within amino acids 5 and 241 of Human PHF17. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| PHF17 | |
| Polyclonal | |
| Unconjugated | |
| Q6IE81 | |
| JADE1 | |
| Recombit fragment corresponding to a region within amino acids 5 and 241 of Human PHF17 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1.02 mg/mL | |
| JADE1 | |
| JADE1, PHD protein Jade-1, gene for apoptosis and differentiation in epithelia, protein Jade-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.