Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543553
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543553 Supplier Invitrogen™ Supplier No. PA543553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RRCIFALAVR VSLPSSPVPA SPASSSGADQ RLPSQSLSSK QKGHTWALET Sequence homology: Human: 100%; Yeast: 92%; Zebrafish: 79%.

PHF19 contains 2 PHD-type zinc fingers. It acts as a transcriptional repressor. Isoform 1 and isoform 2 inhibit transcription from an HSV-tk promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PHF19
Gene Accession No. Q5T6S3
Gene Alias 3110009G19Rik; 3321402G02Rik; DKFZP727G051; hPCL3; MTF2L1; Pcl3; PHD finger protein 19; phf13; Phf19; polycomb like 3; polycomb-like 3; polycomb-like protein 3; si:dkeyp-51b7.1; TDRD19B; tudor domain containing 19B
Gene Symbols PHF19
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PHF19.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26147
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.