Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298813APC
Description
PHF2 Polyclonal antibody specifically detects PHF2 in Human samples. It is validated for Western BlotSpecifications
PHF2 | |
Polyclonal | |
Western Blot | |
GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PHF2. | |
0.1 mL | |
Neuroscience | |
5253 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction