Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552749 Supplier Invitrogen™ Supplier No. PA552749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEIREQTKSK SEAKWKYKNS KPDSLLKMEE EQKLEKSPLA GNKDNKFSFS FSNKKLLGSK ALRPPTSPGV FGALQNFKED KPKPVRDEYE YVSDDGELKI DEFPIRRKKN APKRDLSFLL DKKAVLPTPV TKPKLD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes a protein which contains a zinc finger-like PHD (plant homeodomain) finger, distinct from other classes of zinc finger motifs, and a hydrophobic and highly conserved domain. The PHD finger shows the typical Cys4-His-Cys3 arrangement. PHD finger genes are thought to belong to a diverse group of transcriptional regulators possibly affecting eukaryotic gene expression by influencing chromatin structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF2
Gene Accession No. O75151
Gene Alias CENP-35; centromere protein 35; GRC5; JHDM1E; jumonji C domain-containing histone demethylase 1E; Kiaa0662; Lysine-specific demethylase PHF2; PHD finger protein 2; Phf2
Gene Symbols PHF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF2. Recombinant protein control fragment (Product #RP-89704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.