Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556211 Supplier Invitrogen™ Supplier No. PA556211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRVKPKKKKK KKKKTKPECP CSEEISDTSQ EPSPPKAFAV TRCGSSHKPG VHMSPQLHGP ESGHHKGKVK ALEEDNLSES SSESFLWSDD EYGQDVDVTT NP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Methyllysine-binding protein, component of the MOF histone acetyltransferase protein complex. Not required for maintaining the global histone H4 'Lys-16' acetylation (H4K16ac) levels or locus specific histone acetylation, but instead works downstream in transcriptional regulation of MOF target genes. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Contributes to methyllysine-dependent p53/TP53 stabilization and up-regulation after DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF20
Gene Accession No. Q9BVI0
Gene Alias 6820402O20Rik; C20orf104; GLEA2; Glioma-expressed antigen 2; HCA58; Hepatocellular carcinoma-associated antigen 58; hepatocellular carcinoma-associated antigen 58 homolog; Novel zinc finger protein; NZF; PHD finger 20-like protein; PHD finger protein 20; PHD finger protein 20-like protein; Phf20; TDRD20A; transcription factor TZP; tudor domain containing 20A; TZP
Gene Symbols PHF20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF20. Recombinant protein control fragment (Product #RP-95252).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51230
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.