Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHF20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556212
Description
Immunogen sequence: PSIAPTAVDS NSQTLQPITL ELRRRKISKG CEVPLKRPRL DKNSSQEKSK NYSENTDKDL SRRRSSRLST NGTHEILDPD LVVSDLV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Methyllysine-binding protein, component of the MOF histone acetyltransferase protein complex. Not required for maintaining the global histone H4 'Lys-16' acetylation (H4K16ac) levels or locus specific histone acetylation, but instead works downstream in transcriptional regulation of MOF target genes. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Contributes to methyllysine-dependent p53/TP53 stabilization and up-regulation after DNA damage.
Specifications
| PHF20 | |
| Polyclonal | |
| Unconjugated | |
| PHF20 | |
| 6820402O20Rik; C20orf104; GLEA2; Glioma-expressed antigen 2; HCA58; Hepatocellular carcinoma-associated antigen 58; hepatocellular carcinoma-associated antigen 58 homolog; Novel zinc finger protein; NZF; PHD finger 20-like protein; PHD finger protein 20; PHD finger protein 20-like protein; Phf20; TDRD20A; transcription factor TZP; tudor domain containing 20A; TZP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51230 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BVI0 | |
| PHF20 | |
| Recombinant protein corresponding to Human PHF20. Recombinant protein control fragment (Product #RP-95253). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction