Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF20L1 Monoclonal Antibody (OTI1F4)
GREENER_CHOICE

Catalog No. PIMA527206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527206 Supplier Invitrogen™ Supplier No. MA527206
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PHF20L1 Monoclonal Antibody for Western Blot, IHC (P)

The exact function of PHF20L1 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF20L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHF20L1
Gene Accession No. A8MW92
Gene Alias CGI-72; E130113K22Rik; fi20g04; LOW QUALITY PROTEIN: PHD finger protein 20-like protein 1; PHD finger protein 20 like 1; PHD finger protein 20-like 1; PHD finger protein 20-like protein 1; PHD finger protein 20-like protein 1-like protein; Phf20l1; si:dkey-97i18.2; TDRD20B; tudor domain containing 20B; tudor domain-containing PHF20l1-like protein; tudor domain-containing protein PHF20L1; up-regulated in lung cancer 1; URLC1; wu:fi20g04; wu:fi20g04 protein
Gene Symbols PHF20L1
Host Species Mouse
Immunogen Full length human recombinant protein of PHF20L1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51105
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.