Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF21A Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204043AF532
Description
PHF21A Polyclonal antibody specifically detects PHF21A in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PHF21A | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
BHC80a, BHC80BM-006, BRAF35-HDAC complex protein BHC80, KIAA1696BRAF35/HDAC2 complex (80 kDa), PHD finger protein 21A | |
The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_001095272.1 (GeneID 51317). | |
0.1 mL | |
Neuroscience | |
51317 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction