Learn More
Invitrogen™ PHF21A Polyclonal Antibody

Description
Immunogen sequence: TETTFTFPAP VQPVSLPSPT STDGDIHEDF CSVCRKSGQL LMCDTCSRVY HLDCLDPPLK TIPKGMWICP RCQDQMLKKE EAIPWPGTLA IVHSYIAYKA AKEEEKQKLL KWSSDLKQER EQLEQKVKQL SNSISKCMEM KNTILARQKE MHSSLEKVKQ LIRLIHGIDL SKPVDSEATV GAISNGPDCT PPANAATSTP APSPSSQSCT ANCNQGEETK.
Specifications
Specifications
| Antigen | PHF21A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Phf21a |
| Gene Accession No. | Q6ZPK0, Q96BD5 |
| Gene Alias | 80kDa; Bhc; Bhc80; BHC80a; BM-006; Braf35/HDAC complex (Bhc); BRAF35/HDAC2 complex; BRAF35/HDAC2 complex (80 kDa); BRAF35-HDAC complex protein BHC80; D030065N23Rik; KIAA1696; mBHC80; PFTF1; PHD finger protein 21A; Phf21a; RGD1560612 |
| Gene Symbols | Phf21a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.