Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF21A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PHF21A |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7734
![]() ![]() |
Proteintech Group Inc
121271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PHF21A (PHD finger protein 21A) is a component of the BHC complex, a corepressor complex that represses transcription of neuron-specific genes in non-neuronal cells. The BHC complex is recruited at RE1/NRSE sites by REST and acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier. In the BHC complex, PHF21A may act as a scaffold.Specifications
PHF21A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6ZPK0, Q96BD5 | |
192285, 362166, 51317 | |
PHF21A Fusion Protein Ag2771 | |
Primary | |
-20°C | |
PHF21A |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BHC80, BHC80a, BM 006, KIAA1696, PHD finger protein 21A, PHF21A | |
PHF21A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title