Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF21B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596831 Supplier Invitrogen™ Supplier No. PA596831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRLASNYLNN PLFLTARANE DPCWKNEITH DEHCAACKRG ANLQPCGTCP GAYHLSCLEP PLKTAPKGVW VCPRCQQKAL KKDEGVPWTG MLAIVHSYVT HKTVKEEEKQ KLLQRGSELQ NEHQQLEERD RRLASAVQKC LELKTSLLAR QRGTQSSLDR LRALLRLIQG EQLLQVTMTT TSPAPLLAGP WTKPSVAATH PTVQHPQGHN; Positive Samples: Mouse brain, Mouse spinal cord, Rat brain.

PHF21B (PHD finger protein 21B), also known as PHF4, is a 531 amino acid protein that contains one PHD-type zinc finger and is expressed as multiple alternatively spliced isoforms. The gene encoding PHF21B maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF21B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHF21B
Gene Accession No. Q8C966, Q96EK2
Gene Alias A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739
Gene Symbols PHF21B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 322-531 of human PHF21B (NP_6124241).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112885, 271305, 300117
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.