Learn More
Invitrogen™ PHF21B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144054
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human U20S whole cell. ICC/IF: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
PHF21B (PHD finger protein 21B), also known as PHF4, is a 531 amino acid protein that contains one PHD-type zinc finger and is expressed as multiple alternatively spliced isoforms. The gene encoding PHF21B maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, neurofibromatosis type 2, autism and schizophrenia.
Specifications
PHF21B | |
Polyclonal | |
Unconjugated | |
PHF21B | |
A730032D07Rik; BHC80L; KIAA1661; PHD finger protein 21B; PHD finger protein 4; PHF21B; PHF4; RGD1308739 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112885 | |
-20°C | |
Lyophilized |
ELISA, Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4 mg trehalose and no preservative | |
Q96EK2 | |
PHF21B | |
E.coli-derived human PHF21B recombinant protein (Position: E291-M499). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.