Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562603 Supplier Invitrogen™ Supplier No. PA562603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATVVGGEAP VPVLPTPPEA PRPPATVHPE GVPPADSESK EVGSTETSQD GDASSSEGEM RVMDEDIMVE SGDDSWDLIT CYCRKPF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

Acts as a negative regulator of autophagy, through promoting ubiquitination and degradation of LRSAM1, an E3 ubiquitin ligase that promotes autophagy in response to starvation or infecting bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF23
Gene Accession No. Q9BUL5
Gene Alias FLJ16355; hJUNE-1b; hypothetical protein MGC2941; JUNE-1; MGC2941; novel PHD-finger protein JUNE1; PDH-containing protein JUNE-1; PHD finger protein 23; PHD finger protein 23a; PHD zinc finger containing protein JUNE1; PHF23
Gene Symbols PHF23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF23. Recombinant protein control fragment (Product #RP-93167).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79142
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.