Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569766 Supplier Thermo Scientific Supplier No. PA569766
Add to Cart
Add to Cart

PHF23 Polyclonal Antibody

PHF23 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF23
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PHF23
Gene Accession No. Q8BSN5
Gene Alias FLJ16355; hJUNE-1b; hypothetical protein MGC2941; JUNE-1; MGC2941; novel PHD-finger protein JUNE1; PDH-containing protein JUNE-1; PHD finger protein 23; PHD finger protein 23a; PHD zinc finger containing protein JUNE1; PHF23
Gene Symbols PHF23
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human PHF23 (aa 191-240).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78246
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.