Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555255 Supplier Invitrogen™ Supplier No. PA555255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFTTVLHKQR NKPQQNLQED LPTAVEPLME VTKQEPPKPL RFLPGVLIGW ENQPTTLELA NKPLPVDDIL QSLLGTTGQV YDQAQSVMEQ N Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

This gene encodes a member of a PHD finger-containing gene family. This gene may function as a transcription factor and may be involved in glioblastomas development. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Mar 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF3
Gene Accession No. Q92576
Gene Alias 2310061N19Rik; KIAA0244; mKIAA0244; PHD finger protein 3; Phf3
Gene Symbols PHF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF3. Recombinant protein control fragment (Product #RP-94713).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23469
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.