Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556004 Supplier Invitrogen™ Supplier No. PA556004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKHHPDLIFC RKQAGVAIGR LCEKCDGKCV ICDSYVRPCT LVRICDECNY GSYQGRCVIC GGPGVSDAYY CKECTIQEKD RDGCPKIVNL GSSKTDLFYE RK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Acts as a transcriptional regulator by binding to the GJA1/Cx43 promoter and enhancing its up-regulation by ESR1/ER- alpha. Also involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF5A
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF5A
Gene Accession No. Q7RTV0
Gene Alias 1110007B08Rik; bK223H9.2; INI; PHD finger protein 5A; PHD finger-like domain protein 5A; PHD finger-like domain-containing protein 5A; PHD-finger 5a; Phf5a; Rds3; SAP14b; SF3b14b; SF3B7; splicing factor 3B associated 14 kDa protein; splicing factor 3b, subunit 7; splicing factor 3B-associated 14 kDa protein
Gene Symbols PHF5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF5A. Recombinant protein control fragment (Product #RP-91508).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.