Learn More
Invitrogen™ PHF5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113851
Description
Immunogen sequence: ICGGPGVSDA YYCKECTIQE KDRDGCPKIV NLGSSKTDLF YERKKYGFKK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Acts as a transcriptional regulator by binding to the GJA1/Cx43 promoter and enhancing its up-regulation by ESR1/ER- alpha. Also involved in pre-mRNA splicing.
Specifications
| PHF5A | |
| Polyclonal | |
| Unconjugated | |
| PHF5A | |
| 1110007B08Rik; bK223H9.2; INI; PHD finger protein 5A; PHD finger-like domain protein 5A; PHD finger-like domain-containing protein 5A; PHD-finger 5a; Phf5a; Rds3; SAP14b; SF3b14b; SF3B7; splicing factor 3B associated 14 kDa protein; splicing factor 3b, subunit 7; splicing factor 3B-associated 14 kDa protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7RTV0 | |
| PHF5A | |
| Synthetic peptide directed towards the middle region of human PHF5A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.