Learn More
PHF7 Mouse anti-Human, Clone: OTI1C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505156

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene.Specifications
PHF7 | |
Monoclonal | |
Unconjugated | |
PHF7 | |
HSPC045, HSPC226, NYD-SP6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BWX1 | |
PHF7 | |
Human recombit protein fragment corresponding to amino acids 155-381 of human PHF7 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.