Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557594
Description
Immunogen sequence: KTVKEKKECQ RLRKSAKTRR VTQRKPSSGP VCWLCLREPG DPEKLGEFLQ KDNISVHYFC LILSSKLPQR GQSNRGFHGF LPEDIKKEAA RASRKICFVC Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.
Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene.
Specifications
PHF7 | |
Polyclonal | |
Unconjugated | |
PHF7 | |
1700006H01Rik; 1700010P14Rik; AI427892; AW555949; HSPC045; HSPC226; NYD-SP6; PHD finger protein 7; PHF7; testicular secretory protein Li 34; testis development protein NYD-SP6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BWX1 | |
PHF7 | |
Recombinant protein corresponding to Human PHF7. Recombinant protein control fragment (Product #RP-96248). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction