Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540090
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540090 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540090 Supplier Invitrogen™ Supplier No. PA540090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHF8 Polyclonal Antibody for Western Blot, ELISA

PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4
Gene PHF8
Gene Accession No. Q80TJ7, Q9UPP1
Gene Alias 9830141C09Rik; histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; Kiaa1111; mKIAA1111; MRXSSD; PHD finger protein 8; PHF8; RGD1561065; ZNF422
Gene Symbols PHF8
Host Species Rabbit
Immunogen Three different KLH-conjugated synthetic peptides corresponding to human PHF8 (PHD finger protein 8).
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23133, 320595
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.