Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558537 Supplier Invitrogen™ Supplier No. PA558537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPSTQEAIQ GMLCMANLQS SSSSPATSSL QAWWTGGQDR SSGSSSSGLG TVSNSPASQR TPGKRPIKRP AYWRTESEEE EENASLDEQD SLGACFKDAE YIYPSLESDD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHF8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF8
Gene Accession No. Q9UPP1
Gene Alias 9830141C09Rik; histone lysine demethylase PHF8; JHDM1F; jumonji C domain-containing histone demethylase 1F; Kiaa1111; mKIAA1111; MRXSSD; PHD finger protein 8; PHF8; RGD1561065; ZNF422
Gene Symbols PHF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHF8. Recombinant protein control fragment (Product #RP-95462).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23133
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.