Learn More
PHF8 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00343

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee and Crab-eating ma
PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.Specifications
PHF8 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q80TJ7, Q9UPP1 | |
PHF8 | |
Between 650 and 700 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
PHF8 | |
JHDM1F, MRXSSD, ZNF422, Histone lysine demethylase PHF8, PHD finger protein 8, KIAA1111 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23133, 320595 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.