Learn More
PHF8 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301772A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
PHD finger protein 8 is a jumonji domain containing protein. Like other members of the jumonji family, PHF8 may therefore play a role in histone demethylation. Mutations in PHF8 lead to Siderius type X-linked mental retardation (MRXSSD), a mild to borderline type of mental retardation.Specifications
PHF8 | |
Polyclonal | |
Unconjugated | |
PHF8 | |
JHDM1F, MRXSSD, ZNF422, Histone lysine demethylase PHF8, PHD finger protein 8, KIAA1111 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23133, 320595 | |
4° C | |
Liquid |
ChIP sequencing (ChIP-seq), Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q80TJ7, Q9UPP1 | |
Phf8 | |
Between 650 and 700 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.