Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF8 Rabbit anti-Human, Mouse, Rat, Clone: 2H9D4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PHF8 |
---|---|
Clone | 2H9D4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PHF8 Monoclonal antibody specifically detects PHF8 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PHF8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 1.14.11.27, histone lysine demethylase PHF8, jumonji C domain-containing histone demethylase 1F, KIAA1111ZNF422JHDM1F, MRXSSD, PHD finger protein 8DKFZp686E0868 | |
A synthetic peptide corresponding to a sequence within amino acids 800-900 of human PHF8 (Q9UPP1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
2H9D4 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
23133 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title