Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHGDH Antibody (OTI5C4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273372H
Description
PHGDH Monoclonal specifically detects PHGDH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PHGDH | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PHGDH | |
Full length recombinant protein of human PHGDH (NP_006614) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI5C4 | |
Western Blot, Immunohistochemistry | |
3,3-phosphoglycerate dehydrogenase, 3PGDH, 3-PGDH, D-3-phosphoglycerate dehydrogenase, EC 1.1.1, EC 1.1.1.95, MGC3017, PDG, PGAD, PGD, PGDH, phosphoglycerate dehydrogenase, SERA | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
26227.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction