Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHGDH Monoclonal Antibody (OTI5C4)
GREENER_CHOICE

Catalog No. PIMA526812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526812 Supplier Invitrogen™ Supplier No. MA526812
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PHGDH Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

PHGDH gene encodes the enzyme which is involved in the early steps of L-serine synthesis in animal cells. L-serine is required for D-serine and other amino acid synthesis. The enzyme requires NAD/NADH as a cofactor and forms homotetramers for activity. Mutations in this gene have been found in a family with congenital microcephaly, psychomotor retardation and other symptoms. Multiple alternatively spliced transcript variants have been found, however the full-length nature of most are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHGDH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHGDH
Gene Accession No. O08651, O43175, Q61753
Gene Alias 2-oxoglutarate reductase; 3PGDH; 3-PGDH; 3-phosphoglycerate dehydrogenase; 3-phosphoglycerate dehyrogenase; 4930479N23; A10; D-3-phosphoglycerate dehydrogenase; epididymis secretory protein Li 113; HEL-S-113; Malate dehydrogenase; NLS; NLS1; PDG; PGAD; PGD; PGDH; PGDH3; Phgdh; PHGDHD; phosphoglycerate dehydrogenase; SERA
Gene Symbols PHGDH
Host Species Mouse
Immunogen Full length human recombinant protein of PHGDH produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236539, 26227, 58835
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.