Learn More
PHGDH Mouse anti-Human, Clone: OTI4A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806782

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
3-Phosphoglycerate dehydrogenase (PHGDH) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.Specifications
PHGDH | |
Monoclonal | |
Unconjugated | |
PHGDH | |
3-PGDH, 3PGDH, HEL-S-113, NLS, NLS1, PDG, PGAD, PGD, PGDH, PHGDHD, SERA | |
Mouse | |
Affinity Chromatography | |
RUO | |
26227 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43175 | |
PHGDH | |
Full length human recombit protein of human PHGDH produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.