Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA582863 Supplier Invitrogen™ Supplier No. PA582863
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KVLISDSLDP CCRKILQDGG LQVVEKQNLS KEELIAELQD CEGLIVRSAT KVTADVINAA EKLQVVGRAG TGVDNVDLEA ATRKGILVMN T Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 100%.

PHGDH gene encodes the enzyme which is involved in the early steps of L-serine synthesis in animal cells. L-serine is required for D-serine and other amino acid synthesis. The enzyme requires NAD/NADH as a cofactor and forms homotetramers for activity. Mutations in this gene have been found in a family with congenital microcephaly, psychomotor retardation and other symptoms. Multiple alternatively spliced transcript variants have been found, however the full-length nature of most are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHGDH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHGDH
Gene Accession No. O43175
Gene Alias 2-oxoglutarate reductase; 3PGDH; 3-PGDH; 3-phosphoglycerate dehydrogenase; 3-phosphoglycerate dehyrogenase; 4930479N23; A10; D-3-phosphoglycerate dehydrogenase; epididymis secretory protein Li 113; HEL-S-113; Malate dehydrogenase; NLS; NLS1; PDG; PGAD; PGD; PGDH; PGDH3; Phgdh; PHGDHD; phosphoglycerate dehydrogenase; SERA
Gene Symbols PHGDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHGDH. Recombinant protein control fragment (Product #RP-91731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.