Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PhiKan 083, Tocris Bioscience™

Catalog No. 432610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-261-0 10 mg
43-265-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-261-0 Supplier R&D Systems Supplier No. 4326/10
This item is not returnable. View return policy

p53 stabilizing agent

p53 stabilizing agent; preferentially binds mutated (Y220C) p53 over wild-type p53 at a site distinct from functional DNA/protein interaction regions. Slows rate of thermal denaturation of p53-Y220C.

Chemical Identifiers

CAS 1050480-30-2
Molecular Formula C16H19ClN2
Molecular Weight (g/mol) 274.79
InChI Key IXWVUPURFWFRNE-UHFFFAOYSA-N
Synonym phikan 083 hydrochloride, n-9-ethyl-9h-carbazol-3-yl methyl-n-methylamine hydrochloride, 9-ethylcarbazol-3-yl methyl methyl amine hydrochloride, phikan083, phikan083 hydrochloride hplc, 1-9-ethylcarbazol-3-yl-n-methylmethanamine;hydrochloride, 9-ethyl-n-methyl-9h-carbazole-3-methanamine hydrochloride, 1-9-ethyl-9h-carbazol-3-yl-n-methylmethanamine hydrochloride
PubChem CID 16255105
IUPAC Name [(9-ethyl-9H-carbazol-3-yl)methyl](methyl)amine hydrochloride
SMILES Cl.CCN1C2=C(C=CC=C2)C2=C1C=CC(CNC)=C2

Specifications

Quantity 10 mg
Formula Weight 274.79
Percent Purity >99%
Chemical Name or Material PhiKan 083
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.