Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577167
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PHIP (pleckstrin homology domain interacting protein), also known as ndrp or WDR11, is a 1,821 amino acid protein that contains eight N-terminal WD40 repeats and two bromodomains. It is expressed in skeletal muscle (localizing to the cytosol and nucleus) and primary beta cells (localizing to the nucleus) and acts as a transcriptional activator. PHIP is known to interact with various members of the insulin receptor substrate (IRS) family. The IRS family of proteins mediate insulin receptor signaling and play an important role in insulin-producing beta cell proliferation and survival. PHIP specifically associates with the PH domain of IRS-1 and may function to link IRS-1 to insulin receptors, indicating a vital role of PHIP in the regulation of insulin signaling.
Specifications
PHIP | |
Polyclonal | |
Unconjugated | |
PHIP | |
2810004D21Rik; 4632404O06Rik; BRWD2; DCAF14; DDB1 and CUL4 associated factor 14; DDB1- and CUL4-associated factor 14; FLJ20705; IRS-1 PH domain-binding protein; Ndrp; neuronal differentiation related protein; neuronal differentiation-related protein; PH-interacting protein; PHIP; pleckstrin homology domain interacting protein; WD repeat domain 11; WD repeat-containing protein 11; WDR11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55023, 83946 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8VDD9, Q8WWQ0 | |
PHIP | |
Recombinant full length Human PHIP. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction