Learn More
Invitrogen™ PHIP Polyclonal Antibody

Description
Immunogen sequence: KTILENSVKH SKALNTLSSP GQSSFSHGTR NNSAKENMEK EKPVKRKMKS SVLPKASTLS KSSAVIEQGD CKNNALVPGT IQVNGHGGQP SKLVKRGPGR KPKVEVNTNS GEIIHKKRGR KPKKLQYAKP EDLEQNNVHP IRDEVLPSST CNFLSETNNV KEDLLQKKNR GGRKPKRKMK TQKLDADLLV PASVKVLRRS NRKKIDDPID EEEEFEELKG SEPHMRTRNQ GRRTAFYNED DSEEEQRQLL FEDTSLTFGT SSRGRVRKLT EKAKANLIGW.
Specifications
Specifications
| Antigen | PHIP |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PHIP |
| Gene Accession No. | Q8VDD9, Q8WWQ0 |
| Gene Alias | 2810004D21Rik; 4632404O06Rik; BRWD2; DCAF14; DDB1 and CUL4 associated factor 14; DDB1- and CUL4-associated factor 14; FLJ20705; IRS-1 PH domain-binding protein; Ndrp; neuronal differentiation related protein; neuronal differentiation-related protein; PH-interacting protein; PHIP; pleckstrin homology domain interacting protein; WD repeat domain 11; WD repeat-containing protein 11; WDR11 |
| Gene Symbols | PHIP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.