Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHIP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561656
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-1656 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1656 Supplier Bethyl Laboratories, Inc Supplier No. A302055AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

PHIP (pleckstrin homology domain interacting protein), also known as ndrp or WDR11, is a 1,821 amino acid protein that contains eight N-terminal WD40 repeats and two bromodomains. It is expressed in skeletal muscle (localizing to the cytosol and nucleus) and primary B cells (localizing to the nucleus) and acts as a transcriptional activator. PHIP is known to interact with various members of the insulin receptor substrate (IRS) family. The IRS family of proteins mediate insulin receptor signaling and play an important role in insulin-producing B cell proliferation and survival. PHIP specifically associates with the PH domain of IRS-1 and may function to link IRS-1 to insulin receptors, indicating a vital role of PHIP in the regulation of insulin signaling.

Specifications

Antigen PHIP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PHIP
Gene Accession No. Q8WWQ0
Gene Alias BRWD2, DCAF14, DDB1 and CUL4 associated factor 14, FLJ20705, IRS-1 PH domain-binding protein, ndrp, PHIP, WD repeat-containing protein 11, WDR11
Gene Symbols PHIP
Host Species Rabbit
Immunogen Between 1771 and 1821
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55023
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.