Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKA1/PHKA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599685 Supplier Invitrogen™ Supplier No. PA599685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PHKA1/2.

Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKA1/PHKA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Phka2
Gene Accession No. P18826, P46019, P46020, Q64649, Q8BWJ3
Gene Alias 5330411D17; 6330505C01Rik; 9830108K24Rik; D330034O08; GSD9A; Pcyt1b; PHK; PHKA; Phka1; PHKA2; PhK-alpha-subunit; PHKLA; phosphorylase B kinase alpha subunit; phosphorylase b kinase regulatory subunit alpha liver isoform; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; phosphorylase b kinase regulatory subunit alpha, liver isoform; Phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; phosphorylase kinase; phosphorylase kinase alpha 1; phosphorylase kinase alpha 2; phosphorylase kinase alpha L subunit; Phosphorylase kinase alpha M subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase regulatory subunit alpha 1; phosphorylase kinase regulatory subunit alpha 2; phosphorylase kinase, alpha 1; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis; phosphorylase kinase, alpha 2; phosphorylase kinase, alpha 2 (liver); PYK; PYKL; XLG; XLG2
Gene Symbols PHKA1, Phka2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PHKA1(Accession P46020), corresponding to amino acid residues A42-M92.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110094, 18679, 5255, 5256, 64561, 678739
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.