Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551508 Supplier Invitrogen™ Supplier No. PA551508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHSLRCSAEE ATEGLMNLSP SAMKNLLHHI LSGKEFGVER SVRPTDSNVS PAISIHEIGA VGATKTERTG IMQLKSEIKQ SPGTSMTPSS GSFPSAYDQQ SSKDSRQGQW Q Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 80%.

Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHKA1
Gene Accession No. P46020
Gene Alias 5330411D17; 9830108K24Rik; Pcyt1b; Phka; Phka1; PhK-alpha-subunit; phosphorylase B kinase alpha subunit; phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform; phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform; phosphorylase kinase; phosphorylase kinase alpha 1; phosphorylase kinase alpha M subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase regulatory subunit alpha 1; phosphorylase kinase, alpha 1; phosphorylase kinase, alpha 1 (muscle); phosphorylase kinase, alpha 1 (muscle), muscle glycogenosis
Gene Symbols PHKA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHKA1. Recombinant protein control fragment (Product #RP-91886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.