Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551772 Supplier Invitrogen™ Supplier No. PA551772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GARVKLGNLS EFLTTSFYTY LTFLDPDCDE KLFDNASEGT FSPDSDSDLV GYLEDTCNQE SQDELDHYIN HLLQSTSLRS YLPPLCKNTE DRHVFSAIHS TRDILSVMAK AKGLEVPFVP MTLPTKVLSA HRKS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 36%.

Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKA2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHKA2
Gene Accession No. P46019
Gene Alias 6330505C01Rik; D330034O08; GSD9A; Phk; Phka2; PHKLA; phosphorylase b kinase regulatory subunit alpha liver isoform; phosphorylase b kinase regulatory subunit alpha, liver isoform; phosphorylase kinase; phosphorylase kinase alpha 2; phosphorylase kinase alpha L subunit; phosphorylase kinase alpha-subunit; phosphorylase kinase regulatory subunit alpha 2; phosphorylase kinase, alpha 2; phosphorylase kinase, alpha 2 (liver); PYK; PYKL; RP3-499B10.2; XLG; XLG2
Gene Symbols PHKA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHKA2. Recombinant protein control fragment (Product #RP-92162).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.