Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596901 Supplier Invitrogen™ Supplier No. PA596901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIHIGWIISN NPELFSGMLK IRIGWIIHAM EYELQIRGGD KPALDLYQLS PSEVKQLLLD ILQPQQNGRC WLNRRQIDGS LNRTPTGFYD RVWQILERTP NGIIVAGKHL PQQPTLSDMT MYEMNFSLLV EDTLGNIDQP QYRQIVVELL MVVSIVLERN PELEFQDKVD LDRLVKEAFN EFQKDQSRLK EIEKQDDMTS FYNTPPLGKR GTCSYLTKAV MNLLLEGEVK PNNDDPCLIS; Positive Samples: HepG2, SW620, Jurkat, BT474; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.

Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PHKD. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHKB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHKB
Gene Accession No. Q7TSH2, Q93100
Gene Alias AI463271; Phkb; Phosphorylase B; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase; phosphorylase kinase beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; phosphorylase kinase regulatory subunit beta; phosphorylase kinase subunit beta; phosphorylase kinase, beta
Gene Symbols PHKB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 854-1093 of human PHKB (NP_0002841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102093, 5257
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.