Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHKB Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA537015

Catalog No. PIPA537015


Only null left
Add to Cart

Description

Description

This antibody detects endogenous protein at a molecular weight of 125 kDa. Purity is >95% by SDS-PAGE.

Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PHKD. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PHKB
Polyclonal
Unconjugated
PHKB
AI463271; Phkb; Phosphorylase B; Phosphorylase b kinase regulatory subunit beta; phosphorylase kinase; phosphorylase kinase beta; phosphorylase kinase beta subunit; phosphorylase kinase beta-subunit; phosphorylase kinase regulatory subunit beta; phosphorylase kinase subunit beta; phosphorylase kinase, beta
Rabbit
Antigen affinity chromatography
RUO
102093, 361377, 5257
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q7TSH2, Q93100
PHKB
Synthetic peptide corresponding to amino acids 699-744 of human PHKB.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.